The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145121.1 | NP_001138593.1 | 4 - 5 | 629 | 97 |
XM_017010859.1 | XP_016866348.1 | 4 - 5 | 600 | 97 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004610.3 | NP_004601.3 | 4 - 5 | 629 | 97 |
XM_011536082.2 | XP_011534384.1 | 2 - 3 | 525 | 97 | |
XM_011536084.2 | XP_011534386.1 | 4 - 5 | 624 | 97 | |
XM_011536085.2 | XP_011534387.1 | 4 - 5 | 803 | 97 | |
XM_011536088.2 | XP_011534390.1 | 3 - 4 | 443 | 97 | |
XM_011536090.2 | XP_011534392.1 | 3 - 4 | 390 | 97 | |
XM_017011228.1 | XP_016866717.1 | 3 - 4 | 483 | 97 | |
XM_017011229.1 | XP_016866718.1 | 3 - 4 | 483 | 97 | |
XM_017011230.1 | XP_016866719.1 | 3 - 4 | 483 | 97 | |
XM_017011231.1 | XP_016866720.1 | 3 - 4 | 483 | 97 | |
GenBank mRNA | BC063451.1 | - | 4 - 5 | 585 | 97 |
U03399.1 | - | 4 - 5 | 575 | 97 |