Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM64
Entrez Gene ID
120146
Gene Name
tripartite motif containing 64
Gene Aliases
C11orf28, TRIM64A
Chromosome Location
Chr.11: 89966068 - 89974147 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136486.1 NP_001129958.1 2 - 3 506 189
XM_011542585.2 XP_011540887.1 3 - 4 595 189

Gene Symbol
TRIM64B
Entrez Gene ID
642446
Gene Name
tripartite motif containing 64B
Gene Aliases
-
Chromosome Location
Chr.11: 89870188 - 89878456 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164397.1 NP_001157869.1 2 - 3 506 189
XM_011542954.2 XP_011541256.1 3 - 4 595 189
GenBank mRNA JF968449.1 - 2 - 3 491 189

View Full Product Details