Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDIA3
Entrez Gene ID
2923
Gene Name
protein disulfide isomerase family A member 3
Gene Aliases
ER60, ERp57, ERp60, ERp61, GRP57, GRP58, HEL-S-269, HEL-S-93n, HsT17083, P58, PI-PLC
Chromosome Location
Chr.15: 43746392 - 43772606 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005313.4 NP_005304.3 1 - 1 51 84
GenBank mRNA BG826781.1 - 1 - 1 52 84

View Full Product Details