Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIRLET7G
- Entrez Gene ID
-
406890
- Gene Name
- microRNA let-7g
- Gene Aliases
-
LET7G, MIRNLET7G, hsa-let-7g, let-7g
- Chromosome Location
-
Chr.3: 52268278 - 52268361 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_029660.1 |
|
1 - 1 |
45 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
protein phosphatase, Mg2+/Mn2+ dependent 1M |
PPM1M |
PP2C-eta, PP2CE, PP2Ceta |
-17.7kb
|
Anti-sense |
Chr. - 52245725 - 52250599 on Build GRCh38 |
Downstream |
glycerate kinase |
GLYCTK |
HBEBP2, HBEBP4, HBeAgBP4A |
19.5kb
|
Anti-sense |
Chr. - 52287558 - 52295256 on Build GRCh38 |
View Full Product Details