Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR138-1
- Entrez Gene ID
-
406929
- Gene Name
- microRNA 138-1
- Gene Aliases
-
MIRN138-1, mir-138-1
- Chromosome Location
-
Chr.3: 44114212 - 44114310 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_029700.1 |
|
1 - 1 |
47 |
59 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
abhydrolase domain containing 5 |
ABHD5 |
CDS, CGI58, IECN2, NCIE2 |
-391.5kb
|
Sense |
Chr. - 43690883 - 43722725 on Build GRCh38 |
Downstream |
testis and ovary specific PAZ domain containing 1 |
TOPAZ1 |
C3orf77 |
127.6kb
|
Sense |
Chr. - 44241886 - 44337071 on Build GRCh38 |
View Full Product Details