The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204813.1 | NP_001191742.1 | 3 - 4 | 1310 | 106 |
NM_002526.3 | NP_002517.1 | 3 - 4 | 1310 | 106 | |
GenBank mRNA | AK075008.1 | - | 3 - 4 | 785 | 106 |
AK314661.1 | - | 3 - 4 | 824 | 106 | |
BC065937.1 | - | 3 - 4 | 779 | 106 | |
DQ186653.1 | - | 3 - 4 | 773 | 106 | |
X55740.1 | - | 3 - 4 | 803 | 106 |