The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204878.1 | NP_001191807.1 | 3 - 4 | 1061 | 93 |
NM_004874.3 | NP_004865.1 | 4 - 5 | 1169 | 93 | |
GenBank mRNA | AF095194.2 | - | 4 - 5 | 929 | 93 |
AF111116.1 | - | 4 - 5 | 887 | 93 | |
AK304072.1 | - | 3 - 4 | 838 | 93 | |
AK314020.1 | - | 4 - 5 | 942 | 93 | |
BC000800.1 | - | 3 - 4 | 1061 | 93 | |
BC038505.1 | - | 4 - 5 | 963 | 93 |