Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C16orf89
Entrez Gene ID
146556
Gene Name
chromosome 16 open reading frame 89
Gene Aliases
-
Chromosome Location
Chr.16: 5042772 - 5066145 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098514.2 NP_001091984.2 1 - 2 440 97
NM_152459.4 NP_689672.4 1 - 2 440 97
XM_005255143.3 XP_005255200.2 1 - 2 266 97
XM_017022972.1 XP_016878461.1 1 - 2 266 97
XM_017022973.1 XP_016878462.1 1 - 2 266 97
XM_017022974.1 XP_016878463.1 1 - 2 266 97
GenBank mRNA AK075242.1 - 1 - 2 266 97
AK300708.1 - 1 - 2 214 97
AY358483.1 - 1 - 2 405 97
BC033681.1 - 1 - 2 335 97

View Full Product Details