Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C16orf89
Entrez Gene ID
146556
Gene Name
chromosome 16 open reading frame 89
Gene Aliases
-
Chromosome Location
Chr.16: 5042772 - 5066145 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098514.2 NP_001091984.2 6 - 7 1101 105
NM_152459.4 NP_689672.4 6 - 7 1101 105
XM_005255143.3 XP_005255200.2 6 - 7 927 105
XM_011522392.2 XP_011520694.1 4 - 5 442 105
GenBank mRNA AK075242.1 - 7 - 8 1023 105
AK300708.1 - 6 - 7 875 105
AY358483.1 - 6 - 7 1066 105
BC033681.1 - 6 - 7 996 105

View Full Product Details