Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR345
- Entrez Gene ID
-
442910
- Gene Name
- microRNA 345
- Gene Aliases
-
MIRN345, hsa-mir-345, mir-345
- Chromosome Location
-
Chr.14: 100307859 - 100307956 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_029906.1 |
|
1 - 1 |
49 |
72 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
solute carrier family 25 member 29 |
SLC25A29 |
C14orf69, CACL, ORNT3 |
-1.3kb
|
Anti-sense |
Chr. - 100277986 - 100306547 on Build GRCh38 |
Downstream |
solute carrier family 25 member 47 |
SLC25A47 |
C14orf68, HDMCP, HMFN1655 |
15.4kb
|
Sense |
Chr. - 100323342 - 100330378 on Build GRCh38 |
View Full Product Details