Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SNX17
- Entrez Gene ID
-
9784
- Gene Name
- sorting nexin 17
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 27370496 - 27377533 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_049786.1 |
|
2 - 3 |
385 |
75 |
GenBank mRNA |
AK298856.1 |
- |
2 - 3 |
266 |
75 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BE545019.1 |
- |
2 - 3 |
217 |
|
Chr. 2 - 27370660 - 27374398 [+] on Build GRCh38 |
74 |
GenBankmRNA |
DC337598.1 |
- |
2 - 3 |
266 |
|
Chr. 2 - 27370615 - 27374159 [+] on Build GRCh38 |
75 |
GenBankmRNA |
DC305670.1 |
- |
2 - 3 |
261 |
|
Chr. 2 - 27370620 - 27374111 [+] on Build GRCh38 |
75 |
GenBankmRNA |
DC366374.1 |
- |
2 - 3 |
266 |
|
Chr. 2 - 27370615 - 27373970 [+] on Build GRCh38 |
75 |
GenBankmRNA |
BI761363.1 |
- |
2 - 3 |
254 |
|
Chr. 2 - 27370633 - 27374758 [+] on Build GRCh38 |
75 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
eukaryotic translation initiation factor 2B subunit delta |
EIF2B4 |
EIF-2B, EIF2B, EIF2Bdelta |
-812.0
|
Anti-sense |
Chr. - 27364352 - 27370457 on Build GRCh38 |
Downstream |
zinc finger protein 513 |
ZNF513 |
HMFT0656, RP58 |
4.1kb
|
Anti-sense |
Chr. - 27377231 - 27380866 on Build GRCh38 |
View Full Product Details