Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TRAPPC3
- Entrez Gene ID
-
27095
- Gene Name
- trafficking protein particle complex 3
- Gene Aliases
-
BET3
- Chromosome Location
-
Chr.1: 36136569 - 36156053 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073098.1 |
|
1 - 2 |
177 |
84 |
GenBank mRNA |
AA446176.1 |
- |
1 - 2 |
131 |
84 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA429882.1 |
- |
1 - 2 |
|
|
Chr. 1 - 36137032 - 36149423 [+] on Build GRCh38 |
84 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
collagen type VIII alpha 2 |
COL8A2 |
FECD, FECD1, PPCD, PPCD2 |
-10.8kb
|
Sense |
Chr. - 36095236 - 36126207 on Build GRCh38 |
Downstream |
MAP7 domain containing 1 |
MAP7D1 |
PARCC1, RPRC1 |
6.5kb
|
Anti-sense |
Chr. - 36155965 - 36180849 on Build GRCh38 |
View Full Product Details