The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017370.2 | NP_001017370.1 | 3 - 4 | 705 | 93 |
GenBank mRNA | BX647872.1 | - | 3 - 4 | 595 | 93 |
CR749318.1 | - | 3 - 4 | 626 | 93 |