The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195790.1 | NP_001182719.1 | 2 - 3 | 1201 | 116 |
XM_017024005.1 | XP_016879494.1 | 4 - 5 | 2167 | 116 | |
XM_017024006.1 | XP_016879495.1 | 9 - 10 | 4369 | 116 | |
XM_017024007.1 | XP_016879496.1 | 7 - 8 | 3770 | 116 | |
XM_017024008.1 | XP_016879497.1 | 7 - 8 | 7952 | 116 | |
XM_017024009.1 | XP_016879498.1 | 6 - 7 | 7882 | 116 | |
GenBank mRNA | AK172761.1 | - | 4 - 5 | 2002 | 116 |