The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270894.1 | NP_001257823.1 | 2 - 3 | 278 | 108 |
GenBank mRNA | CD251784.1 | - | 2 - 3 | 296 | 108 |