Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FDX1L
Entrez Gene ID
112812
Gene Name
ferredoxin 1 like
Gene Aliases
FDX2
Chromosome Location
Chr.19: 10310211 - 10316015 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031734.3 NP_001026904.2 3 - 4 330 99
GenBank mRNA AK300568.1 - 3 - 4 323 99
BC063460.1 - 3 - 4 314 99
DA243025.1 - 3 - 4 330 99

View Full Product Details