Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNHIT3
Entrez Gene ID
9326
Gene Name
zinc finger HIT-type containing 3
Gene Aliases
TRIP3
Chromosome Location
Chr.17: 36486627 - 36499310 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281434.1 NP_001268363.1 2 - 3 189 174

View Full Product Details