Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- HECTD2
- Entrez Gene ID
-
143279
- Gene Name
- HECT domain E3 ubiquitin protein ligase 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 91409280 - 91514829 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104291.1 |
|
14 - 15 |
1634 |
126 |
GenBank mRNA |
BX537856.1 |
- |
14 - 15 |
1398 |
126 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB074343.1 |
- |
3 - 4 |
276 |
|
Chr. 10 - 91491251 - 91498184 [+] on Build GRCh38 |
126 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
HECTD2 antisense RNA 1 |
HECTD2-AS1 |
|
|
Anti-sense |
Chr. - 91306962 - 91611460 on Build GRCh38 |
Downstream |
protein phosphatase 1 regulatory subunit 3C |
PPP1R3C |
PPP1R5 |
132.1kb
|
Anti-sense |
Chr. - 91628440 - 91633101 on Build GRCh38 |
View Full Product Details