Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ETV1
- Entrez Gene ID
-
2115
- Gene Name
- ETS variant 1
- Gene Aliases
-
ER81
- Chromosome Location
-
Chr.7: 13891229 - 13991425 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_120445.1 |
|
11 - 12 |
1370 |
126 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ADP ribosylation factor like GTPase 4A |
ARL4A |
ARL4 |
-1.2mb
|
Anti-sense |
Chr. - 12686827 - 12690934 on Build GRCh38 |
Downstream |
diacylglycerol kinase beta |
DGKB |
DAGK2, DGK, DGK-BETA |
235.3kb
|
Sense |
Chr. - 14145049 - 14974777 on Build GRCh38 |
View Full Product Details