Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC101928650
- Entrez Gene ID
-
101928650
- Gene Name
- uncharacterized LOC101928650
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 170461317 - 170532609 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_125958.1 |
|
2 - 3 |
370 |
82 |
GenBank mRNA |
HY035829.1 |
- |
2 - 3 |
370 |
82 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
HY220936.1 |
- |
1 - 2 |
|
|
Chr. 1 - 170461318 - 170462415 [+] on Build GRCh38 |
82 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 1142 |
LINC01142 |
|
-177.1kb
|
Sense |
Chr. - 170271405 - 170284208 on Build GRCh38 |
Downstream |
golgin, RAB6 interacting |
GORAB |
GO, NTKLBP1, SCYL1BP1 |
69.7kb
|
Anti-sense |
Chr. - 170532122 - 170553834 on Build GRCh38 |
View Full Product Details