Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FMOD
- Entrez Gene ID
-
2331
- Gene Name
- fibromodulin
- Gene Aliases
-
FM, SLRR2E
- Chromosome Location
-
Chr.1: 203340621 - 203351429 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_103757.1 |
|
1 - 2 |
397 |
67 |
GenBank mRNA |
DA918421.1 |
- |
1 - 2 |
236 |
67 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG899252.1 |
- |
1 - 2 |
135 |
|
Chr. 1 - 203342183 - 203351086 [-] on Build GRCh38 |
67 |
GenBankmRNA |
BG541058.1 |
- |
|
57 |
|
Chr. - - [-] on Build |
67 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
BTG anti-proliferation factor 2 |
BTG2 |
APRO1, PC3, TIS21 |
-31.0kb
|
Anti-sense |
Chr. - 203305536 - 203309602 on Build GRCh38 |
Downstream |
proline and arginine rich end leucine rich repeat protein |
PRELP |
MST161, MSTP161, SLRR2A |
124.3kb
|
Anti-sense |
Chr. - 203475755 - 203491352 on Build GRCh38 |
View Full Product Details