Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PMP22
- Entrez Gene ID
-
5376
- Gene Name
- peripheral myelin protein 22
- Gene Aliases
-
CMT1A, CMT1E, DSS, GAS-3, GAS3, HMSNIA, HNPP, Sp110
- Chromosome Location
-
Chr.17: 15229777 - 15265357 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104018.1 |
|
1 - 2 |
200 |
161 |
GenBank mRNA |
HY099893.1 |
- |
1 - 2 |
170 |
161 |
KR259962.1 |
- |
1 - 2 |
156 |
161 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG705748.1 |
- |
1 - 2 |
167 |
|
Chr. 17 - 15230753 - 15265319 [-] on Build GRCh38 |
161 |
GenBankmRNA |
BI600130.1 |
- |
1 - 2 |
167 |
|
Chr. 17 - 15230511 - 15265319 [-] on Build GRCh38 |
161 |
GenBankmRNA |
DB502250.1 |
- |
1 - 2 |
163 |
|
Chr. 17 - 15230919 - 15265319 [-] on Build GRCh38 |
161 |
GenBankmRNA |
CD516473.1 |
- |
1 - 2 |
170 |
|
Chr. 17 - 15230579 - 15265300 [-] on Build GRCh38 |
161 |
GenBankmRNA |
HY293418.1 |
- |
2 - 3 |
|
|
Chr. 17 - 15230910 - 15265288 [+] on Build GRCh38 |
161 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 4731 |
MIR4731 |
|
|
Sense |
Chr. - 15251627 - 15251696 on Build GRCh38 |
Downstream |
tektin 3 |
TEKT3 |
|
38.5kb
|
Sense |
Chr. - 15303811 - 15343682 on Build GRCh38 |
View Full Product Details