Product Details

Assay Reference Genome
Location

Chr.13:40791871 on build GRCh38
Cytoband
13q14.11
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
10166
Gene Symbol
SLC25A15
Gene Name
solute carrier family 25 member 15
Gene Aliases
D13S327, HHH, ORC1, ORNT1
Location
Chr.13:40789411-40812460 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
SLC25A15 NM_014252.3 NP_055067.1
AF112968.1 AAD45238.1
AF177333.1 AAG17977.1
AK296862.1
AK313602.1
AL538057.3
BC002702.2 AAH02702.1
BI461478.1
DB505424.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3892332 Chr13:40619338 - 40820761 on Build GRCh38 Gain MIR621 SLC25A15 TPTE2P5 MRPS31 MIR320D1 FOXO1

View Full Product Details