The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253729.1 | NP_001240658.1 | 11 - 12 | 1155 | 201 |
NR_045582.1 | 11 - 12 | 1260 | 201 | ||
GenBank mRNA | AY899849.1 | - | 11 - 12 | 892 | 201 |