Product Details

Assay Reference Genome
Location

Chr.2:218272950 on build GRCh38
Cytoband
2q35
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
25953
Gene Symbol
PNKD
Gene Name
paroxysmal nonkinesigenic dyskinesia
Gene Aliases
BRP17, DYT8, FKSG19, FPD1, KIPP1184, MR-1, MR1, PDC, PKND1, TAHCCP2
Location
Chr.2:218270392-218346793 on build GRCh38
Assay Gene Location
Overlaps Exon 3 - Intron 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
PNKD NM_001077399.2 NP_001070867.1
NM_015488.4 NP_056303.3
XM_017003771.1 XP_016859260.1
AB033010.1 BAA86498.1
AF318057.1 AAL25716.1
AF417001.1 AAL08573.1
AF418285.1 AAL08580.1
AK289867.1
AL080092.1 CAB45707.2
BC021118.1 AAH21118.1
BC036457.2 AAH36457.1
BQ028777.1

View Full Product Details