Gene Transcripts

Gene Symbol
Cfap43
Entrez Gene ID
100048534
Gene Name
cilia and flagella associated protein 43
Gene Aliases
4632415N18Rik, 4930428C11Rik, 4930463G05Rik, AI429486, D19Ertd652e, Wdr96
Chromosome Location
Chr. 19: 47735267 - 47837836 on Build GRCm38
UniGene ID
Mm.29457
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027559.2 NP_081835.2 30 3891
GenBank mRNA BC107402.1 - 7 633
BC107403.1 - 7 633

View Full Product Details