Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029878.3 | NP_084154.1 | 20 | 1989 | |
GenBank mRNA | AK041464.1 | - | 20 | 1937 | |
AK050199.1 | - | 20 | 1992 | ||
AK054343.1 | - | 20 | 1948 | ||
AK212113.1 | - | 1 | 85 | ||
BC059843.1 | - | 20 | 1909 | 6843690 |