Gene Transcripts

Gene Symbol
Ccr2
Entrez Gene ID
12772
Gene Name
chemokine (C-C motif) receptor 2
Gene Aliases
Cc-ckr-2, Ccr2a, Ccr2b, Ckr2, Ckr2a, Ckr2b, Cmkbr2, mJe-r
Chromosome Location
Chr. 9: 124101918 - 124109140 on Build GRCm38
UniGene ID
Mm.6272
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009915.2 NP_034045.1 3 1148
GenBank mRNA AK046579.1 - 1382
AK138599.1 - 2 1461
AK162705.1 - 1 154
BC138803.1 - 2 1108 8861825
BC138804.1 - 2 1108 8861826
U47035.1 - 2 1057
U51717.1 - 2 1106
U56819.1 - 3 1148

View Full Product Details