Gene Transcripts

Gene Symbol
Rasgrf2
Entrez Gene ID
19418
Gene Name
RAS protein-specific guanine nucleotide-releasing factor 2
Gene Aliases
6330417G04Rik, AW048350, Grf2, Ras-GRF2
Chromosome Location
Chr. 13: 91880407 - 91988042 on Build GRCm38
UniGene ID
Mm.248630
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009027.3 NP_033053.2 2567
GenBank mRNA AK158472.1 - 2732
U67326.2 - 2567

View Full Product Details