Gene Transcripts

Gene Symbol
Slc12a2
Entrez Gene ID
20496
Gene Name
solute carrier family 12, member 2
Gene Aliases
9330166H04Rik, Nkcc1, mBSC2, sy-ns
Chromosome Location
Chr. 18: 57878513 - 57946821 on Build GRCm38
UniGene ID
Mm.399997
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009194.3 NP_033220.2 5 1257
GenBank mRNA U13174.1 - 5 1254

View Full Product Details