Gene Transcripts
- Gene Symbol
- Itga10
- Entrez Gene ID
- 213119
- Gene Name
- integrin, alpha 10
- Gene Aliases
- A630048L14
- Chromosome Location
- Chr. 3: 96645584 - 96664519 on Build GRCm38
- UniGene ID
- Mm.32741
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK037222.1 |
- |
4 |
221 |
|
BC115770.1 |
- |
24 |
2889 |
40085898 |
View Full Product Details