Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013854.1 | NP_038882.1 | 14 | 1469 | |
GenBank mRNA | AK145549.1 | - | 5 | 621 | |
BC040783.1 | - | 3 | 321 | ||
BC046965.1 | - | 14 | 1440 | 6485365 | |
BC063094.1 | - | 14 | 1469 | 6516514 |