Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178691.4 | NP_848806.2 | 2 | 1075 | |
GenBank mRNA | AK036938.1 | - | 2 | 1076 | |
AK036991.1 | - | 2 | 1077 | ||
AK044678.1 | - | 2 | 871 | ||
AK053730.1 | - | 2 | 865 | ||
BC099948.1 | - | 2 | 868 | 30097649 | |
BC139034.1 | - | 2 | 1001 | 8862056 |