Gene Transcripts

Gene Symbol
Col6a4
Entrez Gene ID
68553
Gene Name
collagen, type VI, alpha 4
Gene Aliases
1110001D15Rik, AI413310, AU023415, Dvwa, EG235580, Vwa6
Chromosome Location
Chr. 9: 105990318 - 106096691 on Build GRCm38
UniGene ID
Mm.28854
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026763.2 NP_081039.2 5 1829
GenBank mRNA AB370265.1 - 5 1921
AM231151.1 - 5 1829

View Full Product Details