Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001003717.1 | NP_001003717.1 | 15 | 2136 | |
NM_175489.3 | NP_780698.2 | 16 | 2173 | ||
GenBank mRNA | AK173174.1 | - | 4 | 334 | |
BC141383.1 | - | 16 | 1872 | 9055996 | |
CF745582.1 | - | 3 | 215 |