Gene Transcripts

Gene Symbol
Galm
Entrez Gene ID
319625
Gene Name
galactose mutarotase
Gene Aliases
A530057M15Rik, AU015645, AU020959
Chromosome Location
Chr. 17: 80127458 - 80185032 on Build GRCm38
UniGene ID
Mm.29098
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176963.4 NP_795937.1 2 530
GenBank mRNA AK040998.1 - 2 531
AK085153.1 - 2 331
AK161687.1 - 2 329
AK170014.1 - 2 329
BC028818.1 - 2 333 5135570

View Full Product Details