Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025579.3 | NP_079855.2 | 4 | 524 | |
GenBank mRNA | AK007811.1 | - | 4 | 487 | |
AK013133.1 | - | 4 | 487 | ||
AK132927.1 | - | 4 | 525 | ||
AK139748.1 | - | 4 | 441 | ||
BC019668.1 | - | 4 | 469 | 3493047 |