Gene Transcripts

Gene Symbol
Pbrm1
Entrez Gene ID
66923
Gene Name
polybromo 1
Gene Aliases
2610016F04Rik, AI507524, BAF180, Pb1
Chromosome Location
Chr. 14: 31019138 - 31121592 on Build GRCm38
UniGene ID
Mm.27913
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081251.1 NP_001074720.1 27 4299
GenBank mRNA AK009120.1 - 3 325
BC055708.1 - 3 407

View Full Product Details