Gene Transcripts

Gene Symbol
Pef1
Entrez Gene ID
67898
Gene Name
penta-EF hand domain containing 1
Gene Aliases
2600002E23Rik, Peflin
Chromosome Location
Chr. 4: 130107556 - 130128134 on Build GRCm38
UniGene ID
Mm.20818
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026441.4 NP_080717.2 3 511
GenBank mRNA AK007394.1 - 3 453
AK013238.1 - 3 453
AK046247.1 - 3 453
AK075978.1 - 3 516

View Full Product Details