Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001321.3 | NP_001001321.2 | 9 | 692 | |
GenBank mRNA | AB117931.1 | - | 9 | 673 | |
AB117932.1 | - | 9 | 673 | ||
AK154257.1 | - | 9 | 718 | ||
AK159148.1 | - | 9 | 693 | ||
BC046402.1 | - | 3 | 232 | ||
BC107210.1 | - | 8 | 639 | ||
BC107211.1 | - | 9 | 672 | 40051417 |