Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028198.2 | NP_082474.1 | 23, 24 | 2742 | |
GenBank mRNA | AF343581.1 | - | 24, 25 | 2742 | |
AK122486.1 | - | 846 | |||
BC039187.1 | - | 1667 | |||
BC092371.1 | - | 2814 | |||
BC131660.1 | - | 23, 24 | 2738 | 40110670 | |
BC131661.1 | - | 23, 24 | 2738 | 40110672 |