Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030026.2 | NP_084302.1 | 16 | 1642 | |
GenBank mRNA | AK132265.1 | - | 16 | 1622 | |
AK143233.1 | - | 16 | 1643 | ||
AK145564.1 | - | 16 | 1594 | ||
AK146844.1 | - | 16 | 1602 | ||
AK146865.1 | - | 16 | 1592 | ||
BC033050.1 | - | 3 | 304 | ||
BC040098.1 | - | 8 | 654 | ||
BC051500.1 | - | 4 | 461 | ||
BC058987.1 | - | 8 | 656 | ||
BC141193.1 | - | 16 | 1608 | 9055805 | |
BC141194.1 | - | 16 | 1608 | 9055806 |