Gene Transcripts

Gene Symbol
Akp3
Entrez Gene ID
11648
Gene Name
alkaline phosphatase 3, intestine, not Mn requiring
Gene Aliases
Akp-3, IAP, dIAP
Chromosome Location
Chr. 1: 87125008 - 87127912 on Build GRCm38
UniGene ID
Mm.377076
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007432.2 NP_031458.2 5 605
GenBank mRNA BC115745.1 - 5 604
BC145627.1 - 3 801

View Full Product Details