Gene Transcripts

Gene Symbol
Phox2b
Entrez Gene ID
18935
Gene Name
paired-like homeobox 2b
Gene Aliases
Dilp1, NBPhox, Pmx2b
Chromosome Location
Chr. 5: 67094397 - 67099126 on Build GRCm38
UniGene ID
Mm.62505
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008888.3 NP_032914.1 1 207
GenBank mRNA AK052579.2 - 1 138
BC079610.1 - 1 207 30360139
Y14493.1 - 1 240

View Full Product Details