Gene Transcripts

Gene Symbol
Shisa2
Entrez Gene ID
219134
Gene Name
shisa family member 2
Gene Aliases
9430059P22Rik, MAd2, Tmem46, mShisa, shisa
Chromosome Location
Chr. 14: 59625281 - 59631660 on Build GRCm38
UniGene ID
Mm.275409
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145463.5 NP_663438.1 2 1209
GenBank mRNA AB280737.1 - 2 963
AK034905.1 - 2 1184
AK087512.1 - 2 1179

View Full Product Details