Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009453.3 | NP_033479.2 | 2, 3 | 472 | |
NM_178794.4 | NP_848909.2 | 2, 3 | 472 | ||
GenBank mRNA | AK030296.1 | - | 2, 3 | 404 | |
AK045504.1 | - | 2, 3 | 404 | ||
AK078070.1 | - | 2, 3 | 117 | ||
BC065693.1 | - | 2, 3 | 388 | ||
BC083160.1 | - | 2, 3 | 365 | ||
BC099617.1 | - | 2, 3 | 370 | ||
D45205.1 | - | 2, 3 | 395 |