Gene Transcripts

Gene Symbol
Prss42
Entrez Gene ID
235628
Gene Name
protease, serine 42
Gene Aliases
Tessp2
Chromosome Location
Chr. 9: 110797261 - 110803744 on Build GRCm38
UniGene ID
Mm.292285
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153099.1 NP_694739.1 2, 3 372
GenBank mRNA AB052292.1 - 2, 3 372
AK133402.1 - 2, 3 237
BC060984.1 - 2, 3 391 6774705

View Full Product Details