Gene Transcripts

Gene Symbol
Nol6
Entrez Gene ID
230082
Gene Name
nucleolar protein family 6 (RNA-associated)
Gene Aliases
AA410091, Nrap
Chromosome Location
Chr. 4: 41114427 - 41124339 on Build GRCm38
UniGene ID
Mm.285797
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139236.3 NP_631982.2 2 355
GenBank mRNA AF361077.1 - 2 304
AF361078.1 - 2 304
AK053117.1 - 2 472
AK089138.1 - 2 275
AK155818.1 - 2 294
AK166559.1 - 2 281
AK166650.1 - 2 281
AK171163.1 - 2 292
AK171421.1 - 2 282
AK195706.1 - 2 211
AK197974.1 - 2 211
AK198310.1 - 2 211
AK199838.1 - 2 212
AK202668.1 - 2 211
AK204352.1 - 2 214
AK209500.1 - 2 211
BC059820.1 - 2 273 6827596
BC060189.1 - 2 283
CN459855.1 - 2 355

View Full Product Details