Gene Transcripts

Gene Symbol
Cga
Entrez Gene ID
12640
Gene Name
glycoprotein hormones, alpha subunit
Gene Aliases
CG-alpha, FSHA, GPHA1, GPHalpha, HCG, LHA, Tsha, aGSU, alpha-GSU, alphaGSU, alphaSU
Chromosome Location
Chr. 4: 34893779 - 34907374 on Build GRCm38
UniGene ID
Mm.1361
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009889.2 NP_034019.1 2 174
GenBank mRNA AF307151.1 - 1 88
BC087926.1 - 2 172 30251641
J00643.1 - 2 142

View Full Product Details