Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138951.1 | NP_620401.1 | 1 | 204 | |
GenBank mRNA | AK149384.1 | - | 1 | 235 | |
AK165811.1 | - | 1 | 102 | ||
BC021608.1 | - | 1 | 116 | 5099426 | |
BC021613.1 | - | 1 | 210 | 5102393 | |
BC024432.1 | - | 1 | 204 | 4972715 | |
BC026825.1 | - | 1 | 151 | 4240199 |